Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mei4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175213.3 | NP_780422.1 | 4 | 1026 | ||
GenBank mRNA | AK043929.1 | - | 4 | 1027 | ||
AK082395.1 | - | 4 | 1066 | |||
AK166209.1 | - | 3 | 816 | |||
BC096613.1 | - | 4 | 1068 | 6469435 |