Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191051.1 | NP_001177980.1 | 2 | 513 | ||
NM_175215.4 | NP_780424.1 | 2 | 176 | |||
GenBank mRNA | AK015716.1 | - | 177 | |||
AK033681.1 | - | 2 | 113 | |||
AK039294.1 | - | 2 | 107 | |||
AK143241.1 | - | 2 | 514 | |||
AK172203.1 | - | 2 | 142 | |||
BC030310.1 | - | 2 | 119 | |||
BC057459.1 | - | 1 | 97 | 4504057 | ||
BC094328.1 | - | 2 | 500 |