Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc2hc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029172.1 | NP_083448.1 | 2 | 111 | ||
GenBank mRNA | AK015837.1 | - | 2 | 80 | ||
BC137685.1 | - | 2 | 111 | 8860704 |