Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081293.1 | NP_001074762.1 | 3 | 571 | ||
GenBank mRNA | AK012860.1 | - | 3 | 446 | ||
AK015972.1 | - | 3 | 482 | |||
AK083674.1 | - | 3 | 394 | |||
AK087157.1 | - | 3 | 397 | |||
AK129150.1 | - | 3 | 250 | |||
AK154692.1 | - | 3 | 377 | |||
BC067054.1 | - | 3 | 376 | 30542928 |