Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029182.1 | NP_083458.1 | 2 | 184 | ||
GenBank mRNA | AK015898.1 | - | 2 | 415 | ||
BC036988.1 | - | 2 | 195 |