Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029196.1 | NP_083472.1 | 2 | 117 | ||
GenBank mRNA | AK016041.1 | - | 2 | 102 | ||
BC119474.1 | - | 1 | 483 | |||
BC152479.1 | - | 1 | 483 | |||
CF982527.1 | - | 2 | 117 | |||
FJ468007.1 | - | 1 | 70 |