Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meiob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029197.1 | NP_083473.1 | 10 | 846 | ||
GenBank mRNA | AK015924.1 | - | 2 | 87 | ||
BC119085.1 | - | 2 | 145 | 8733834 | ||
BC119087.1 | - | 2 | 145 | 8733836 |