Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282419.2 | NP_001269348.1 | 2, 3 | 240 | ||
NM_001282420.2 | NP_001269349.1 | 3, 4 | 302 | |||
NM_001282421.2 | NP_001269350.1 | 3, 4 | 293 | |||
NM_001291304.1 | NP_001278233.1 | 5, 6 | 479 | |||
NM_005000.4 | NP_004991.1 | 3, 4 | 285 | |||
NR_104168.2 | 4, 5 | 494 | ||||
NR_111925.1 | 3, 4 | 283 | ||||
NR_111926.1 | 6, 7 | 443 | ||||
GenBank mRNA | AK315458.1 | - | 3, 4 | 196 | ||
AV713284.1 | - | 2, 3 | 136 | |||
BC000813.1 | - | 3, 4 | 201 | 3453236 | ||
BC020821.1 | - | 3, 4 | 185 | 4701912 | ||
BC070236.1 | - | 3, 4 | 186 | 4290594 | ||
BC070237.1 | - | 3, 4 | 185 | 4659819 | ||
BF670375.1 | - | 436 | ||||
BF790429.1 | - | 6, 7 | 412 | |||
BG400546.1 | - | 4, 5 | 396 | |||
BT006695.1 | - | 3, 4 | 176 | |||
CR933664.1 | - | 3, 4 | 302 | |||
U53468.1 | - | 3, 4 | 199 | |||
U64028.1 | - | 3, 4 | 194 | |||
W44870.1 | - | 3, 4 | 302 |