Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029211.2 | NP_083487.2 | 4, 5 | 410 | ||
GenBank mRNA | AK016037.1 | - | 4, 5 | 641 | ||
AK034806.1 | - | 4, 5 | 1724 | |||
AK078509.1 | - | 4, 5 | 371 | |||
AK159032.1 | - | 4, 5 | 547 | |||
AK159260.1 | - | 4, 5 | 437 | |||
AK168821.1 | - | 4, 5 | 423 | |||
AK169089.1 | - | 4, 5 | 423 | |||
BC022686.1 | - | 4, 5 | 410 | 4241984 |