Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002490.3 | NP_002481.2 | 2 | 338 | ||
GenBank mRNA | AF047182.1 | - | 2 | 202 | ||
AK291874.1 | - | 2 | 216 | |||
AK316562.1 | - | 2 | 212 | |||
BC002772.1 | - | 2 | 199 | 3619356 | ||
CR456529.1 | - | 2 | 338 | |||
CR456714.1 | - | 2 | 198 | |||
CR541997.1 | - | 2 | 198 |