Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002490.3 | NP_002481.2 | 3 | 418 | ||
GenBank mRNA | AF047182.1 | - | 3 | 282 | ||
AK291874.1 | - | 3 | 296 | |||
AK316562.1 | - | 3 | 292 | |||
BC002772.1 | - | 3 | 279 | 3619356 | ||
CR456529.1 | - | 3 | 418 | |||
CR456714.1 | - | 3 | 278 | |||
CR541997.1 | - | 3 | 278 |