Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005001.4 | NP_004992.2 | 3 | 207 | ||
NR_135539.1 | 3 | 207 | ||||
GenBank mRNA | AF050637.1 | - | 3 | 164 | ||
AF054178.1 | - | 3 | 178 | |||
BC003102.1 | - | 3 | 190 | 3537231 | ||
BC107892.1 | - | 3 | 178 | |||
CD172691.1 | - | 3 | 240 | |||
CR456915.1 | - | 3 | 167 | |||
CR542266.1 | - | 3 | 167 | |||
CR975510.1 | - | 3 | 152 | |||
DA189271.1 | - | 3 | 185 |