Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171004.1 | NP_001164475.1 | 11 | 2197 | ||
NM_001171005.1 | NP_001164476.1 | 10 | 1393 | |||
NM_029239.3 | NP_083515.2 | 11 | 2197 | |||
GenBank mRNA | AK030058.1 | - | 10 | 1390 | ||
BC027320.1 | - | 10 | 1596 | |||
BC037012.1 | - | 10 | 1841 | 5342428 | ||
BC089164.1 | - | 11 | 2096 | 5400250 |