Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318195.1 | NP_001305124.1 | 2, 3 | 354 | ||
NM_014222.2 | NP_055037.1 | 2, 3 | 354 | |||
GenBank mRNA | AF044953.1 | - | 2, 3 | 279 | ||
AK301520.1 | - | 2, 3 | 322 | |||
AK314135.1 | - | 2, 3 | 314 | |||
BC001016.2 | - | 2, 3 | 258 | 3345138 | ||
BU602971.1 | - | 2, 3 | 356 |