Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023773.2 | NP_076262.2 | 3 | 1051 | ||
GenBank mRNA | AB041598.2 | - | 3 | 1052 | ||
AK145518.1 | - | 3 | 1016 | |||
AK158773.1 | - | 3 | 1029 | |||
AK159495.1 | - | 3 | 1029 | |||
BC022759.1 | - | 2 | 638 | |||
BC079558.1 | - | 3 | 984 | |||
BC145164.1 | - | 3 | 976 | |||
BC145700.1 | - | 3 | 976 | 40131003 |