Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144539.5 | NP_653122.2 | 6 | 1271 | ||
GenBank mRNA | AB196816.1 | - | 5 | 818 | ||
AB196817.1 | - | 5 | 818 | |||
AB196818.1 | - | 5 | 818 | |||
AB196819.1 | - | 5 | 818 | |||
AF467909.1 | - | 5 | 818 | |||
AF467910.1 | - | 5 | 818 | |||
AK016183.1 | - | 6 | 1070 | |||
AK030135.1 | - | 6 | 1132 | |||
AK030148.1 | - | 6 | 1272 | |||
AK040678.1 | - | 5 | 899 | |||
AK089525.1 | - | 5 | 899 | |||
AK172665.1 | - | 5 | 865 |