Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001444.2 | NP_001001444.1 | 2 | 149 | ||
GenBank mRNA | AF515626.1 | - | 2 | 150 | ||
AK002707.1 | - | 2 | 150 | |||
BC146010.1 | - | 2 | 147 | 40131313 | ||
BC146012.1 | - | 2 | 147 | 40131315 |