Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081123.1 | NP_001074592.1 | 5 | 843 | ||
GenBank mRNA | AB073967.1 | - | 5 | 710 | ||
AK048167.1 | - | 5 | 732 | |||
BC090672.1 | - | 5 | 814 | 30941825 | ||
BC145645.1 | - | 5 | 809 | 40130948 |