Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029278.2 | NP_083554.2 | 1 | 182 | ||
GenBank mRNA | AK011648.1 | - | 1 | 158 | ||
AK079609.1 | - | 1 | 184 | |||
AK148605.1 | - | 1 | 141 | |||
BC024998.1 | - | 1 | 103 | 5012070 | ||
BC043043.1 | - | 1 | 125 | 5701430 |