Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029278.2 | NP_083554.2 | 5 | 790 | ||
GenBank mRNA | AK011648.1 | - | 5 | 766 | ||
AK148605.1 | - | 5 | 749 | |||
AK161728.1 | - | 5 | 728 | |||
BC024998.1 | - | 5 | 711 | 5012070 | ||
BC043043.1 | - | 5 | 733 | 5701430 |