Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029282.1 | NP_083558.1 | 1 | 2104 | ||
GenBank mRNA | AF332067.1 | - | 1 | 2087 | ||
AF332068.1 | - | 1 | 2087 | |||
AK128940.1 | - | 2 | 2154 | |||
BC053067.1 | - | 1 | 2104 | 5698252 |