Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099.4 | NP_001090.2 | 5, 6 | 638 | ||
NM_001134194.1 | NP_001127666.1 | 5, 6 | 638 | |||
XM_011512946.1 | XP_011511248.1 | 5, 6 | 602 | |||
GenBank mRNA | BC007460.1 | - | 5, 6 | 598 | 3951204 | |
BC008493.1 | - | 5, 6 | 595 | 4272289 | ||
BC016344.1 | - | 5, 6 | 602 | 3950907 | ||
BC017877.1 | - | 5, 6 | 588 | |||
M24902.1 | - | 5, 6 | 571 | |||
M34840.1 | - | 5, 6 | 562 | |||
X52174.1 | - | 5, 6 | 571 | |||
X53605.1 | - | 5, 6 | 584 |