Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou5f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029315.1 | NP_083591.1 | 1 | 679 | ||
GenBank mRNA | AK005948.1 | - | 1 | 680 | ||
AK076753.1 | - | 1 | 670 |