Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700020N15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029316.1 | NP_083592.1 | 2 | 334 | ||
GenBank mRNA | AK006180.1 | - | 2 | 316 | ||
BC049765.1 | - | 312 | ||||
CA464409.1 | - | 2 | 336 |
Gene Symbol: Gm6812 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098842.2 | NP_001092312.1 | 2 | 315 |