Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029332.1 | NP_083608.1 | 5 | 858 | ||
GenBank mRNA | AK172125.1 | - | 5 | 791 |