Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199957.1 | NP_001186886.1 | 4 | 350 | ||
NM_001199958.1 | NP_001186887.1 | 5 | 399 | |||
NM_002492.3 | NP_002483.1 | 6 | 506 | |||
GenBank mRNA | AF047181.1 | - | 6 | 483 | ||
AK313822.1 | - | 6 | 484 | |||
AW955118.1 | - | 5 | 356 | |||
BC005271.1 | - | 6 | 497 | 3997377 | ||
BC009796.1 | - | 6 | 501 | 4284776 | ||
BC017909.1 | - | 6 | 496 | |||
BC093071.1 | - | 4 | 327 | 30325282 | ||
BG776403.1 | - | 6 | 513 | |||
BX648034.1 | - | 4 | 344 | |||
DA112751.1 | - | 6 | 509 |