Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029336.1 | NP_083612.1 | 2 | 264 | ||
GenBank mRNA | AK006255.1 | - | 2 | 268 | ||
CA466331.1 | - | 2 | 270 |