Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhej1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029342.4 | NP_083618.3 | 418 | |||
GenBank mRNA | AK006481.1 | - | 412 | |||
BC006063.1 | - | 392 | 3591824 | |||
BC107252.1 | - | 337 | 40052353 | |||
BC107253.1 | - | 337 | 40052354 | |||
BC132359.1 | - | 308 | 40130636 | |||
BC132361.1 | - | 308 | 40130638 | |||
BY095373.1 | - | 341 |