Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acyp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029344.3 | NP_083620.1 | 4 | 280 | ||
GenBank mRNA | AK009134.1 | - | 4 | 280 | ||
AK046238.1 | - | 4 | 289 |