Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029348.2 | NP_083624.2 | 3 | 599 | ||
GenBank mRNA | AK009517.1 | - | 3 | 520 | ||
AK147212.1 | - | 3 | 553 | |||
AK173196.1 | - | 1 | 353 | |||
BC112421.1 | - | 2 | 395 | |||
BC113187.1 | - | 2 | 395 |