Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284367.1 | NP_001271296.1 | 4 | 419 | ||
NM_001284368.1 | NP_001271297.1 | 4 | 328 | |||
NM_005004.3 | NP_004995.1 | 4 | 419 | |||
GenBank mRNA | AF044958.1 | - | 4 | 369 | ||
AF077028.1 | - | 4 | 366 | |||
AF115968.1 | - | 4 | 375 | |||
AK289579.1 | - | 4 | 360 | |||
AL080056.1 | - | 4 | 379 | |||
BC000466.2 | - | 4 | 360 | 2822781 | ||
BC019276.1 | - | 4 | 360 | 2822781 | ||
BM008944.1 | - | 4 | 316 | |||
BM686677.1 | - | 4 | 367 | |||
CF127298.1 | - | 4 | 358 | |||
CR533490.1 | - | 4 | 347 | |||
DA428446.1 | - | 4 | 419 |