Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152895.2 | NP_690855.2 | 23 | 3923 | ||
GenBank mRNA | AK041304.1 | - | 23 | 3930 | ||
AK081306.1 | - | 1 | 2250 | |||
AK220451.1 | - | 23 | 3909 | |||
AY082429.1 | - | 23 | 3906 | |||
AY082430.1 | - | 23 | 3906 | |||
BC019446.1 | - | 2 | 442 | |||
BC034189.1 | - | 10 | 1800 | |||
BC048180.1 | - | 23 | 3906 | 6826125 | ||
BC057318.1 | - | 23 | 3906 | 6826125 |