Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290311.1 | NP_001277240.1 | 17 | 4174 | ||
NM_001290313.1 | NP_001277242.1 | 17 | 4174 | |||
NM_029361.4 | NP_083637.2 | 17 | 4174 | |||
GenBank mRNA | AK129439.1 | - | 6 | 1031 | ||
AK147550.1 | - | 17 | 3861 | |||
AK166741.1 | - | 1 | 1586 | |||
BC046464.1 | - | 10 | 1973 | |||
BC060187.1 | - | 6 | 1205 |