Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278646.1 | NP_001265575.1 | 1, 2 | 193 | ||
NM_005005.2 | NP_004996.1 | 1, 2 | 195 | |||
GenBank mRNA | AF044956.1 | - | 1, 2 | 189 | ||
AF067168.1 | - | 1, 2 | 189 | |||
AF261090.1 | - | 1, 2 | 194 | |||
AI907018.1 | - | 1, 2 | 212 | |||
AK026538.1 | - | 1, 2 | 178 | |||
AK302286.1 | - | 1, 2 | 157 | |||
AK313432.1 | - | 1, 2 | 157 | |||
AL136714.1 | - | 1, 2 | 165 | |||
BC007672.2 | - | 1, 2 | 162 | 3160562 | ||
BG703246.1 | - | 1, 2 | 122 | |||
BQ685604.1 | - | 1, 2 | 145 |