Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023790.2 | NP_076279.1 | 4, 5 | 550 | ||
GenBank mRNA | AF053628.1 | - | 4, 5 | 538 | ||
AK006536.1 | - | 4, 5 | 450 | |||
AK154345.1 | - | 4, 5 | 541 | |||
AK202399.1 | - | 3, 4 | 165 | |||
BC083332.1 | - | 5, 6 | 455 | 6819838 |