Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080808.1 | NP_001074277.1 | 6 | 1277 | ||
GenBank mRNA | AK036757.1 | - | 3 | 737 | ||
BC024804.1 | - | 6 | 1270 | |||
BC126948.1 | - | 6 | 1277 | 40090576 | ||
GU973876.1 | - | 6 | 1277 |