Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029385.2 | NP_083661.2 | 3 | 561 | ||
GenBank mRNA | AK009737.1 | - | 3 | 562 | ||
AK012920.1 | - | 2 | 614 | |||
AK013493.1 | - | 1 | 1397 | |||
BC064048.1 | - | 3 | 548 | 574013 |