Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145625.3 | NP_663600.2 | 8 | 1140 | ||
GenBank mRNA | AK045250.1 | - | 8 | 984 | ||
AK077423.1 | - | 8 | 991 | |||
AK148012.1 | - | 8 | 990 | |||
AK161693.1 | - | 8 | 973 | |||
AK163266.1 | - | 8 | 990 | |||
AK166363.1 | - | 8 | 992 | |||
AK168350.1 | - | 8 | 986 | |||
AK168425.1 | - | 8 | 986 | |||
AK170256.1 | - | 8 | 991 | |||
AK170354.1 | - | 8 | 973 | |||
AK181426.1 | - | 1 | 107 | |||
AK181558.1 | - | 2 | 200 | |||
AK187833.1 | - | 1 | 62 | |||
AK193699.1 | - | 1 | 62 | |||
AK201687.1 | - | 1 | 62 | |||
AK219622.1 | - | 1 | 62 | |||
BC007171.1 | - | 7 | 939 | 3492114 | ||
BC036293.1 | - | 8 | 976 | |||
BC147746.1 | - | 8 | 1049 | 9056695 | ||
BC147748.1 | - | 8 | 1049 | 9056697 |