Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167748.1 | NP_001161220.1 | 10 | 1534 | ||
NM_029412.1 | NP_083688.1 | 12 | 1705 | |||
GenBank mRNA | AK018073.1 | - | 12 | 1780 | ||
AK032017.1 | - | 12 | 1549 |