Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029427.1 | NP_083703.1 | 4 | 664 | ||
GenBank mRNA | AK014771.1 | - | 4 | 665 | ||
BC109360.2 | - | 4 | 664 | 40058461 | ||
BC109361.2 | - | 4 | 664 | 40058463 |