Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166102.1 | NP_001159574.1 | 6 | 1021 | ||
NM_007103.3 | NP_009034.2 | 6 | 1048 | |||
GenBank mRNA | AF053070.1 | - | 6 | 964 | ||
AF092131.1 | - | 6 | 974 | |||
AK223061.1 | - | 6 | 942 | |||
AK293522.1 | - | 5 | 859 | |||
AK300726.1 | - | 4 | 555 | |||
AK316122.1 | - | 6 | 889 | |||
BC007619.1 | - | 3 | 565 | |||
BC008146.1 | - | 6 | 933 | 4156629 | ||
BC015645.2 | - | 6 | 928 | 4648781 | ||
CR456739.1 | - | 6 | 895 | |||
S67973.1 | - | 4 | 637 |