Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318051.1 | NP_001304980.1 | 2, 3 | 215 | ||
NM_002495.3 | NP_002486.1 | 2, 3 | 215 | |||
NR_134475.1 | 3, 4 | 339 | ||||
GenBank mRNA | AF020351.1 | - | 2, 3 | 193 | ||
AK307973.1 | - | 2, 3 | 193 | |||
BC005270.1 | - | 2, 3 | 209 | 3681312 | ||
BU145342.1 | - | 2, 3 | 189 | |||
BU954271.1 | - | 2, 3 | 207 | |||
CD692133.1 | - | 3, 4 | 355 | |||
DB489661.1 | - | 2, 3 | 214 | |||
KU178226.1 | - | 2, 3 | 185 |