Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167946.2 | NP_001161418.1 | 5 | 662 | ||
NM_001286391.1 | NP_001273320.1 | 5 | 662 | |||
NM_001286393.1 | NP_001273322.1 | 4 | 546 | |||
NM_029468.3 | NP_083744.2 | 4 | 682 | |||
GenBank mRNA | AK136168.1 | - | 5 | 589 | ||
AK155121.1 | - | 5 | 361 | |||
AK165387.1 | - | 5 | 586 | |||
AK166399.1 | - | 4 | 546 | |||
BC032193.1 | - | 4 | 437 | |||
BC059014.1 | - | 4 | 613 | 6414779 | ||
BC059844.1 | - | 2 | 662 | |||
CF169872.1 | - | 4 | 257 |