Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167946.2 | NP_001161418.1 | 9 | 1712 | ||
NM_001286391.1 | NP_001273320.1 | 9 | 1731 | |||
NM_001286393.1 | NP_001273322.1 | 8 | 1615 | |||
NM_029468.3 | NP_083744.2 | 8 | 1732 | |||
GenBank mRNA | AK016232.1 | - | 1 | 110 | ||
AK136168.1 | - | 9 | 1639 | |||
AK155121.1 | - | 9 | 1430 | |||
AK166399.1 | - | 8 | 1615 | |||
BC023180.1 | - | 3 | 772 | |||
BC032193.1 | - | 8 | 1506 | |||
BC059014.1 | - | 8 | 1663 | 6414779 | ||
BC059844.1 | - | 6 | 1731 |