Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081428.1 | NP_001074897.1 | 1, 2 | 201 | ||
GenBank mRNA | AK015334.1 | - | 1, 2 | 202 |