Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029491.2 | NP_083767.2 | 15 | 2094 | ||
NM_177038.3 | NP_796012.2 | 15 | 2094 | |||
GenBank mRNA | AK041615.1 | - | 4 | 2181 | ||
AK083747.1 | - | 15 | 2094 | |||
AK173085.1 | - | 8 | 871 | |||
BC048382.1 | - | 4 | 381 |