Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cant1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025617.2 | NP_001020788.1 | 3 | 623 | ||
NM_001025618.2 | NP_001020789.1 | 3 | 581 | |||
NM_001267591.1 | NP_001254520.1 | 2 | 424 | |||
NM_001267592.1 | NP_001254521.1 | 3 | 623 | |||
NM_029502.3 | NP_083778.2 | 3 | 775 | |||
GenBank mRNA | AK006565.1 | - | 3 | 474 | ||
AK017942.1 | - | 3 | 493 | |||
AK049879.1 | - | 3 | 517 | |||
AK085059.1 | - | 2 | 1871 | |||
BC020003.1 | - | 2 | 423 | 4208801 | ||
CD562087.1 | - | 3 | 499 |