Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002496.3 | NP_002487.1 | 4 | 257 | ||
XM_005274013.1 | XP_005274070.1 | 4 | 354 | |||
XM_005274014.2 | XP_005274071.1 | 4 | 381 | |||
XM_011545053.2 | XP_011543355.1 | 4 | 292 | |||
GenBank mRNA | AK002110.1 | - | 3 | 747 | ||
AK302449.1 | - | 4 | 257 | |||
AK314546.1 | - | 4 | 182 | |||
BC119754.2 | - | 4 | 186 | 40119290 | ||
DB498707.1 | - | 4 | 243 | |||
U65579.1 | - | 4 | 180 |