Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ganc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172672.2 | NP_766260.2 | 14, 15 | 2769 | ||
GenBank mRNA | AK076333.1 | - | 13, 14 | 1713 | ||
BC079548.1 | - | 14, 15 | 2231 |