Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021074.4 | NP_066552.2 | 5 | 541 | ||
XM_017025782.1 | XP_016881271.1 | 5 | 731 | |||
GenBank mRNA | AF147367.1 | - | 1 | 16 | ||
AK291865.1 | - | 5 | 437 | |||
AK312129.1 | - | 5 | 459 | |||
BC001632.1 | - | 5 | 451 | 3536345 | ||
BC017487.1 | - | 5 | 449 | 3537815 | ||
BC071689.1 | - | 5 | 431 | 4372795 | ||
BG742948.1 | - | 5 | 541 | |||
CR456928.1 | - | 5 | 427 | |||
M22538.1 | - | 5 | 445 |